Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentadecanoic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1002-84-2
C15H30O2
242.4
MFCD00002745
WQEPLUUGTLDZJY-UHFFFAOYSA-N
pentadecylic acid,n-pentadecanoic acid,pentadecyclic acid,n-pentadecylic acid,unii-ccw02d961f,pentadecyclate,n-pentadecylate,n-pentadecanoate,n-pentadecanoicacid,pentadecansäure
13849
CHEBI:42504
pentadecanoic acid
CCCCCCCCCCCCCCC(=O)O
1002-84-2
C15H30O2
242.4
MFCD00002745
WQEPLUUGTLDZJY-UHFFFAOYSA-N
pentadecylic acid,n-pentadecanoic acid,pentadecyclic acid,n-pentadecylic acid,unii-ccw02d961f,pentadecyclate,n-pentadecylate,n-pentadecanoate,n-pentadecanoicacid,pentadecansäure
13849
CHEBI:42504
pentadecanoic acid
CCCCCCCCCCCCCCC(=O)O
Melting Point 51°C to 53°C
Boiling Point 339°C
Flash Point >110°C
Infrared Spectrum Authentic
Color White
Packaging Glass bottle
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3(CH2)13CO2H
Beilstein 02, 369
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 242.4
Physical Form Powder or Flakes
Percent Purity ≥98.5%
Chemical Name or Material n-Pentadecanoic acid, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 213-693-1

RTECSNumber : RZ1925000

TSCA : TSCA