Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
n-Pentane, HPLC Grade, 99% min
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
Description
n-Pentane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -130°C |
Color | Colorless |
Appearance | Colorless liquid |
Density | 0.626 g/cm3 |
Boiling Point | 36°C |
Quantity | 4 x 4 L |
Flash Point | -49°C (-57°F) |
Ignition Point | 260°C (500°F) |
Physical Form | Liquid |
Refractive Index | 1.358 |
Show More |
Safety and Handling
Hazard Category | H225-H304-H336 |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c |
DOTInformation | Hazard Class: 3, Packing Group: II |
EINECSNumber | 203-692-4 |
RTECSNumber | RZ9450000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction