Learn More
n-Pentane, capillary GC grade, 98+%, Thermo Scientific Chemicals
Description
For organic residue analysis.n-Pentane is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -130°C |
Color | Colorless |
Density | 0.626 g/mL |
Boiling Point | 36°C |
Quantity | 4 x 4 L |
Flash Point | −49°C (57°F) |
Physical Form | Liquid |
Refractive Index | 1.358 |
UN Number | UN1265 |
Merck Index | 14,7116 |
Show More |
Safety and Handling
Hazard Category | H225-H304-H336 |
Hazard Statement | GHS H Statement H225-H304-H336 Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PENTANES |
EINECSNumber | 203-692-4 |
RTECSNumber | QO9500000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.