Learn More
Thermo Scientific™ N-PER™ Neuronal Protein Extraction Reagent PROMO
Description
Neuron cell lysates prepared with the N-PER Reagent may be used for enzymatic activity assays (e.g., Phosphatase, Kinase, ATPase Assays), immunoassays (e.g., Western blots, ELISAs, RIAs) and protein purification. The N-PER Lysis Reagent enables protein extraction to be completed in less than 30 minutes and requires mechanical disruption of tissue (e.g., Dounce Homogenization, Polytron). Typical neuronal protein yields are 70-90μg of protein per mg of brain tissue or 300μg of total protein from 106 primary neuronal cells.
Highlights:
Optimized – efficient extraction of total neuronal protein, including membrane proteins, from tissue or primary cultured cells
Gentle – preserves protein function without compromising yield
Versatile – can be supplemented with protease inhibitors, reducing or chelating agents or required cofactors
Compatible – extracts are suitable for use with total protein, enzymatic and immunological assays and protein purification methods
Recommended for:
Isolate functional proteins from neuronal tissues or primary neuronal cells; Study enzymatic activity assays specific to neuronal tissue; Generate lysates from neuronal tissues for biomarker analysis
Specifications
Specifications
| Quantity | 100 mL |
| Format | Liquid |
| Product Type | Neuronal Protein Extraction Reagent |
| Volume (Metric) | 100 mL |
| Content And Storage | Upon receipt store product at 4°C. |
| Product Line | NE-PER |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.