Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenyl-1-naphthylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
250g
Packaging:
Plastic bottle
90-30-2
C16H13N
219.29
MFCD00003878
XQVWYOYUZDUNRW-UHFFFAOYSA-N
n-phenyl-1-naphthylamine,1-anilinonaphthalene,pana,antioxidant pan,n-1-naphthyl aniline,neozone a,1-naphthalenamine, n-phenyl,phenylnaphthylamine,nonox a,aceto pan
7013
CHEBI:34876
N-phenylnaphthalen-1-amine
C1=CC=C(C=C1)NC2=CC=CC3=CC=CC=C32
90-30-2
C16H13N
219.29
MFCD00003878
XQVWYOYUZDUNRW-UHFFFAOYSA-N
n-phenyl-1-naphthylamine,1-anilinonaphthalene,pana,antioxidant pan,n-1-naphthyl aniline,neozone a,1-naphthalenamine, n-phenyl,phenylnaphthylamine,nonox a,aceto pan
7013
CHEBI:34876
N-phenylnaphthalen-1-amine
C1=CC=C(C=C1)NC2=CC=CC3=CC=CC=C32
Boiling Point 226.0°C (15.0 mmHg)
Packaging Plastic bottle
Infrared Spectrum Authentic
Quantity 25g
Color Brown to Pink
Flash Point >200°C
Melting Point 58.0°C to 62.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula C10H7NHC6H5
Beilstein 12, 1224
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 219.29
Physical Form Crystals or Flakes
Percent Purity 98%
Chemical Name or Material N-Phenyl-1-naphthylamine, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-983-0

RTECSNumber : QM4500000

TSCA : TSCA