Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenylanthranilic acid, 98%

Catalog No. p-3737180
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Plastic bottle

CAS: 91-40-7 | C13H11NO2 | 213.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-40-7
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.24
MDL Number MFCD00002421
InChI Key ZWJINEZUASEZBH-UHFFFAOYSA-N
Synonym n-phenylanthranilic acid, 2-phenylamino benzoic acid, fenamic acid, diphenylamine-2-carboxylic acid, phenylanthranilic acid, 2-carboxydiphenylamine, benzoic acid, 2-phenylamino, o-anilinobenzoic acid, n-phenyl-o-aminobenzoic acid, n-phenyl-2-aminobenzoic acid
PubChem CID 4386
ChEBI CHEBI:34756
IUPAC Name 2-anilinobenzoic acid
SMILES C1=CC=C(C=C1)NC2=CC=CC=C2C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 182°C to 187°C
Color Gray-Green
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula 2-(C6H5NH)C6H4CO2H
Quantity 10 g
Beilstein 14, 327
Merck Index 15, 7384
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot alcohol,slightly soluble in hot benzene and ether
Formula Weight 213.24
Percent Purity 98%
Physical Form Fine Powder
Chemical Name or Material N-Phenylanthranilic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-066-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.