Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenylglycine 97.0+%, TCI America™
SDP

Catalog No. P0180500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0180500G 500 g
P018025G 25 g
P0180100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0180500G Supplier TCI America Supplier No. P0180500G

Chemical Identifiers

CAS 103-01-5
Molecular Formula C8H8NNaO2
Molecular Weight (g/mol) 173.15
MDL Number MFCD00014009
InChI Key YEMGQZDWLLBIEY-UHFFFAOYSA-M
Synonym n-phenylglycine, anilinoacetic acid, 2-phenylamino acetic acid, glycine, n-phenyl, glycine, phenyl, n-phenylglycin, n-phenylaminoacetic acid, phenylamino acetic acid, h-phenylgly-oh, n-phenyl glycine
PubChem CID 66025
ChEBI CHEBI:55477
IUPAC Name sodium 2-(phenylamino)acetate
SMILES [Na+].[O-]C(=O)CNC1=CC=CC=C1

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 500 g
Formula Weight 151.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Phenylglycine
EINECSNumber (3)-1441
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.