Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenylmaleimide, 97%

Catalog No. AC130710250
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 941-69-5 | C10H7NO2 | 173.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 941-69-5
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00005502
InChI Key HIDBROSJWZYGSZ-UHFFFAOYSA-N
Synonym n-phenylmaleimide, maleanil, maleinanil, maleimidobenzene, 1h-pyrrole-2,5-dione, 1-phenyl, 1-phenyl-1h-pyrrole-2,5-dione, maleimide, n-phenyl, 1-phenyl-2,5-dihydro-1h-pyrrole-2,5-dione, unii-9u9kt462vw, n-fenylimid kyseliny maleinove
PubChem CID 13662
IUPAC Name 1-phenylpyrrole-2,5-dione
SMILES C1=CC=C(C=C1)N2C(=O)C=CC2=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 86°C to 90°C
Color Yellow
Boiling Point 162°C to 163°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 21, 400
Fieser 01,845; 13,317
Merck Index 15, 7410
Formula Weight 173.17
Percent Purity 97%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material N-Phenylmaleimide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy
EINECSNumber 213-382-0
RTECSNumber ON5950000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.