Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenylthiobenzamide 98.0+%, TCI America™
SDP

Catalog No. P023325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P023325G 25 g
P02335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P023325G Supplier TCI America Supplier No. P023325G

Chemical Identifiers

CAS 636-04-4
Molecular Formula C13H11NS
Molecular Weight (g/mol) 213.298
MDL Number MFCD00022147
InChI Key BOQKCADLPNLYCZ-UHFFFAOYSA-N
Synonym Thiobenzanilide
PubChem CID 668263
IUPAC Name N-phenylbenzenecarbothioamide
SMILES C1=CC=C(C=C1)C(=S)NC2=CC=CC=C2

Specifications

Melting Point 102°C
Color Yellow
Quantity 25 g
Formula Weight 213.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Phenylthiobenzamide
RTECSNumber CV8925000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.