Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenylthiourea, 97%

Catalog No. aal0669030
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 103-85-5 | C7H8N2S | 152.22 g/mol

N-phenylthiourea is used in paternity testing prior to the advent of DNA testing. It inhibits melanogenesis and is used to grow transparent fish. It is also used as a repellent for rat, rabbits, weasels and also in medical genetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-phenylthiourea is used in paternity testing prior to the advent of DNA testing. It inhibits melanogenesis and is used to grow transparent fish. It is also used as a repellent for rat, rabbits, weasels and also in medical genetics.

Solubility
Soluble in hot water and alcohol.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-85-5
Molecular Formula C7H8N2S
Molecular Weight (g/mol) 152.22
MDL Number MFCD00004933
InChI Key FULZLIGZKMKICU-UHFFFAOYSA-N
Synonym n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea
PubChem CID 676454
ChEBI CHEBI:46261
IUPAC Name phenylthiourea
SMILES NC(=S)NC1=CC=CC=C1

Specifications

Melting Point 149°C to 153°C
Odor Odorless
Quantity 250 g
UN Number UN2811
Beilstein 907309
Merck Index 14,7315
Solubility Information Soluble in hot water and alcohol.
Formula Weight 152.21
Percent Purity 97%
Chemical Name or Material N-Phenylthiourea
Hazard Category H300-H317
Hazard Statement GHS H Statement
H300
Fatal if swallowed.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P310-P302+P352-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 203-151-2
RTECSNumber YU1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.