Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Propargylmaleimide 98.0+%, TCI America™
SDP

Catalog No. P2139100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2139100MG 100 mg
P213925MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2139100MG Supplier TCI America Supplier No. P2139100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 209395-32-4
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.122
MDL Number MFCD12090919
InChI Key OBYJFWVFCFYKNY-UHFFFAOYSA-N
PubChem CID 10214393
IUPAC Name 1-prop-2-ynylpyrrole-2,5-dione
SMILES C#CCN1C(=O)C=CC1=O

Specifications

Melting Point 58°C
Color White-Yellow
Boiling Point 88°C
Quantity 100 mg
Formula Weight 135.12
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Propargylmaleimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.