Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
n-Propyl 4-hydroxybenzoate, 99+%
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
n-Propyl 4-hydroxybenzoate is used as antimicrobial preservatives in pharmaceuticals, foods, cosmetics and shampoos. It is widely used as preservatives in personal care products like creams, lotions and bath products. Further, it is used as a food additive. Its derivative, sodium propyl p-hydroxybenzoate is used as an anti-fungal preservation agent.
Solubility
Soluble in propylene glycol, ethanol, ether, acetone, methanol anddimethyl sulfoxide. Slightly soluble in glycerin. Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong bases and alkali metals.
n-Propyl 4-hydroxybenzoate is used as antimicrobial preservatives in pharmaceuticals, foods, cosmetics and shampoos. It is widely used as preservatives in personal care products like creams, lotions and bath products. Further, it is used as a food additive. Its derivative, sodium propyl p-hydroxybenzoate is used as an anti-fungal preservation agent.
Solubility
Soluble in propylene glycol, ethanol, ether, acetone, methanol anddimethyl sulfoxide. Slightly soluble in glycerin. Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong bases and alkali metals.
Specifications
Specifications
| Melting Point | 95°C to 99°C |
| Flash Point | 180°C (356°F) |
| Quantity | 1000 g |
| Beilstein | 1103245 |
| Merck Index | 14,7866 |
| Solubility Information | Soluble in propylene glycol,ethanol,ether,acetone,methanol anddimethyl sulfoxide. Slightly soluble in glycerin. Insoluble in water. |
| Formula Weight | 180.2 |
| Percent Purity | ≥99% |
| Chemical Name or Material | n-Propyl 4-hydroxybenzoate |
Safety and Handling
| Hazard Category | H315-H320-H373 |
| Precautionary Statement | P262-P264b-P280-P302+P352-P305+P351+P338-P314-P332+P313-P362-P501c |
| EINECSNumber | 202-307-7 |
| RTECSNumber | DH2800000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction