Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl oxalate, 99%

Catalog No. AC159180025
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum

CAS: 95-92-1 | C6H10O4 | 146.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-92-1
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD00009119
InChI Key WYACBZDAHNBPPB-UHFFFAOYSA-N
Synonym ethyl oxalate, diethyl ethanedioate, ethanedioic acid, diethyl ester, oxalic acid, diethyl ester, oxalic ether, diethyloxalate, ethyl oxalate van, oxalic acid diethyl ester, unii-860m3zwf6j, diethylester kyseliny stavelove
PubChem CID 7268
IUPAC Name diethyl oxalate
SMILES CCOC(=O)C(=O)OCC

Specifications

Acidity 0.01% max. (as oxalic acid)
Melting Point -41.0°C
Density 1.0700g/mL
Boiling Point 185.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4085 to 1.4105
Linear Formula C2H5O2CCO2C2H5
Quantity 2.5 kg
Beilstein 02, 535
Fieser 01,250; 02,132; 07,106; 15,133
Specific Gravity 1.07
Merck Index 15, 3142
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,2-propanol and,acetone
Formula Weight 146.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl oxalate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
Wear eye protection/face protection.
EINECSNumber 202-464-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.