Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Butylaniline, 97%

Catalog No. AC150655000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 104-13-2 | C10H15N | 149.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-13-2
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.23
InChI Key OGIQUQKNJJTLSZ-UHFFFAOYSA-N
Synonym 4-n-butylaniline, benzenamine, 4-butyl, p-n-butylaniline, p-butylaniline, 4-butylbenzenamine, p-aminobutylbenzene, p-butylaminobenzene, 1-amino-4-butylbenzene, p-butyl aniline, 4-butylphenylamine
PubChem CID 7694
IUPAC Name 4-butylaniline
SMILES CCCCC1=CC=C(C=C1)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown to Yellow
Density 0.9450g/mL
Boiling Point 133.0°C to 134.0°C (14.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.535
Linear Formula CH3(CH2)3C6H4NH2
Quantity 500 g
Beilstein 12,I,503
Specific Gravity 0.945
Formula Weight 149.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Butylaniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 203-177-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.