Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Propyl Gallate, MP Biomedicals™

Catalog No. ICN10274790 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10274790 500 g
MP210274780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10274790 Supplier MP Biomedicals, Inc Supplier No. 0210274790
Add to Cart
Add to Cart

Off-White Powder

  • Anti-fade reagent in fluorescence microscopy to reduce photobleaching of fluorescent probes such as rhodamine and fluorescein
  • An ester formed by the condensation of gallic acid and propanol
  • This antioxidant has been added to foods containing oils and fats to prevent oxidation
  • An antioxidant that exhibits antimicrobial activity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-79-9
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.2
MDL Number MFCD00002196
InChI Key ZTHYODDOHIVTJV-UHFFFAOYSA-N
PubChem CID 4947
ChEBI CHEBI:10607
IUPAC Name propyl 3,4,5-trihydroxybenzoate
SMILES CCCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 140°C to 150°C
Color Off-white
Loss on Drying 0.0026
Quantity 500 g
Beilstein 1877976
Synonym propyl gallate, n-propyl gallate, progallin p, tenox pg, nipagallin p, gallic acid, propyl ester, gallic acid propyl ester, nipa 49, benzoic acid, 3,4,5-trihydroxy-, propyl ester, 3,4,5-trihydroxybenzene-1-propylcarboxylate
Solubility Information Soluble in ethanol (50mg/mL).
Formula Weight 212.2
Percent Purity 0.992
Physical Form Powder
Chemical Name or Material n-Propyl gallate, Molecular Biology
Grade 0.992
Show More Show Less
EINECSNumber 204-498-2
RTECSNumber LW8400000
storageNote1 Store at Room Temperature (15-30°C).
Recommended Storage Store at Room Temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.