Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Propylbenzene, 98%

Catalog No. aab2146822
Click to view available options
Quantity:
100 g

CAS: 103-65-1 | C9H12 | 120.20 g/mol

n-Propylbenzene is used to prepare benzoic acid. It is employed as a solvent for gas chromatography. It is also used as a solvent and an intermediate in organic synthesis. Further, it is used in fuels and fuel additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Propylbenzene is used to prepare benzoic acid. It is employed as a solvent for gas chromatography. It is also used as a solvent and an intermediate in organic synthesis. Further, it is used in fuels and fuel additives.

Solubility
Miscible with alcohol, ether, acetone, benzene, and petroleum ether. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-65-1
Molecular Formula C9H12
Molecular Weight (g/mol) 120.20
MDL Number MFCD00009377
InChI Key ODLMAHJVESYWTB-UHFFFAOYSA-N
Synonym n-propylbenzene, 1-phenylpropane, phenylpropane, benzene, propyl, 1-propylbenzene, isocumene, n-propyl benzene, propyl-benzene, unii-0wr86zhg2z, propylbenzene, n
PubChem CID 7668
ChEBI CHEBI:42630
IUPAC Name propylbenzene
SMILES CCCC1=CC=CC=C1

Specifications

Melting Point °C to 100°C
Density 0.862
Boiling Point 159°C to 160°C
Flash Point 47°C (116°F)
Assay Percent Range 0.98
Refractive Index 1.491
Quantity 100 g
UN Number UN1993
Beilstein 1903006
Merck Index 14,7844
Solubility Information Miscible with alcohol,ether,acetone,benzene,and petroleum ether. Slightly miscible with water.
Formula Weight 120.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material n-Propylbenzene
Show More Show Less
Hazard Category H226-H304-H335
Hazard Statement GHS H Statement
H304-H226-H335
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 203-132-9
RTECSNumber DA8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.