Learn More
n-Propylbenzene, 98%
Description
n-Propylbenzene is used to prepare benzoic acid. It is employed as a solvent for gas chromatography. It is also used as a solvent and an intermediate in organic synthesis. Further, it is used in fuels and fuel additives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
n-Propylbenzene is used to prepare benzoic acid. It is employed as a solvent for gas chromatography. It is also used as a solvent and an intermediate in organic synthesis. Further, it is used in fuels and fuel additives.
Solubility
Miscible with alcohol, ether, acetone, benzene, and petroleum ether. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | °C to 100°C |
| Density | 0.862 |
| Boiling Point | 159°C to 160°C |
| Flash Point | 47°C (116°F) |
| Assay Percent Range | 0.98 |
| Refractive Index | 1.491 |
| Quantity | 100 g |
| UN Number | UN1993 |
| Beilstein | 1903006 |
| Merck Index | 14,7844 |
| Show More |
Safety and Handling
| Hazard Category | H226-H304-H335 |
| Hazard Statement | GHS H Statement H304-H226-H335 |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 203-132-9 |
| RTECSNumber | DA8750000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.