Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ N-Ras Monoclonal Antibody (06)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA529452
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human NRAS/N-Ras.
NRAS (GTPase NRas) is a membrane protein that shuttles between the Golgi apparatus and the plasma membrane. This shuttling is regulated through palmitoylation and depalmitoylation by the ZDHHC9-GOLGA7 complex. This protein, which has intrinsic GTPase activity, is activated to a GTP-bound form by a GTPase activating protein and inactivated to a GDP-bound form by a guanine nucleotide-exchange factor. Defects in the gene encoding this protein are a cause of juvenile myelomonocytic leukemia (JMML) and Noonan syndrome 6.
Specifications
N-Ras | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P01111 | |
NRAS | |
Recombinant Human NRAS/N-Ras Protein (Met1-Cys186). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
06 | |
Unconjugated | |
NRAS | |
ALPS4; AV095280; CMNS; GTPase NRas; HRAS1; NCMS; Neuroblastoma RAS; neuroblastoma ras oncogene; neuroblastoma RAS viral (v-ras) oncogene homolog; neuroblastoma RAS viral oncogene homolog; Nras; N-ras; N-ras protein part 4; NRAS1; NS6; RAS; RASN; transforming protein N-Ras; v-ras neuroblastoma RAS viral oncogene homolog | |
Mouse | |
Protein A | |
RUO | |
4893 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction