Learn More
Invitrogen™ N-terminal Arginylation Monoclonal Antibody (4A9), PE
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA545373
Description
A 1:1,000 dilution of MA5-45373 was sufficient for detection of N-terminal Arginylation in 0.5 μg of N-terminal Arginine peptide conjugated to BSA by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Specific for N-terminal arginine next to glutamic acid. Does not detect N-terminal arginine next to aspartic acid or internal arginine residues.
Protein arginylation is the post-translational addition of arginine to proteins by arginyltransferase ATE1. Arginylation of proteins has been found to play an essential role in physiological pathways during embryogenesis and adulthood. Arginylation has also been shown to regulate cell stress responses, including ER stress, cytosolic misfolded proteins, and heat stress.
Specifications
N-terminal Arginylation | |
Monoclonal | |
1 mg/mL | |
2.48 mM MES, 95.64 mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Chemical | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
4A9 | |
PE | |
N terminal Arginine; N-terminal Arginine | |
Synthetic N-terminal arginylated peptide conjugated to KLH. | |
100 μg | |
Primary | |
4°C | |
Liquid | |
Chemical |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.