Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ N-terminal Arginylation Monoclonal Antibody (4A9), PerCP
GREENER_CHOICE

Catalog No. PIMA545372
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545372 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545372 Supplier Invitrogen™ Supplier No. MA545372
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-45372 was sufficient for detection of N-terminal Arginylation in 0.5 μg of N-terminal Arginine peptide conjugated to BSA by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Specific for N-terminal arginine next to glutamic acid. Does not detect N-terminal arginine next to aspartic acid or internal arginine residues.

Protein arginylation is the post-translational addition of arginine to proteins by arginyltransferase ATE1. Arginylation of proteins has been found to play an essential role in physiological pathways during embryogenesis and adulthood. Arginylation has also been shown to regulate cell stress responses, including ER stress, cytosolic misfolded proteins, and heat stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen N-terminal Arginylation
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 4A9
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene Alias Arginyltransferase; Arginyltransferase 2; Arginyl-tRNA--protein transferase; Arginyl-tRNA--protein transferase 2; arg-tRNA-protein transferase; arg-tRNA-protein transferase 2; ATE1; ATE2; N terminal Arginine; N-terminal Arginine; R-transferase; R-transferase 2
Host Species Mouse
Immunogen Synthetic N-terminal arginylated peptide conjugated to KLH.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.