Learn More
Invitrogen™ N-terminal Arginylation Monoclonal Antibody (4A9)

Description
MA5-27563 is specific for N-terminal arginine next to glutamic acid. It does not detect N-terminal arginine next to aspartic acid or internal arginine residues. A 1:1,000 dilution of MA5-27563 was sufficient for detection of N-terminal Arginylation in 0.5 μg of N-terminal Arginine peptide conjugated to BSA by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Specific for N-terminal arginine next to glutamic acid. Does not detect N-terminal arginine next to aspartic acid or internal arginine residues.
Specifications
Specifications
| Antigen | N-terminal Arginylation |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 4A9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 |
| Gene Alias | Arginyltransferase; Arginyltransferase 2; Arginyl-tRNA--protein transferase; Arginyl-tRNA--protein transferase 2; arg-tRNA-protein transferase; arg-tRNA-protein transferase 2; ATE1; ATE2; N terminal Arginine; N-terminal Arginine; R-transferase; R-transferase 2 |
| Host Species | Mouse |
| Immunogen | Synthetic N-terminal arginylated peptide conjugated to KLH. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.