Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-4-aminobutyric Acid 98.0+%, TCI America™
SDP

Catalog No. B429125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B429125G 25 g
B42915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B429125G Supplier TCI America Supplier No. B429125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57294-38-9
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD00037313
InChI Key HIDJWBGOQFTDLU-UHFFFAOYSA-N
Synonym boc-gaba-oh, n-boc-gamma-aminobutyric acid, boc-gamma-abu-oh, 4-tert-butoxycarbonyl amino butanoic acid, 4-boc-amino butyric acid, 4-tert-butoxycarbonylaminobutyric acid, 4-tert-butoxycarbonylamino butyric acid, boc-gaba, 4-tert-butoxycarbonylamino butanoic acid
PubChem CID 294894
IUPAC Name 4-{[(tert-butoxy)carbonyl]amino}butanoic acid
SMILES CC(C)(C)OC(=O)NCCCC(O)=O

Specifications

Melting Point 58°C
Boiling Point 82°C
Color White-Yellow
Quantity 25 g
Formula Weight 203.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-4-aminobutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.