Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-4-bromo-D-phenylalanine 98.0+%, TCI America™
SDP

Catalog No. B39451G
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 79561-82-3
Molecular Formula C14H18BrNO4
Molecular Weight (g/mol) 344.205
MDL Number MFCD00270359
InChI Key ULNOXUAEIPUJMK-LLVKDONJSA-N
Synonym boc-d-4-bromophenylalanine, boc-4-bromo-d-phenylalanine, boc-d-phe 4-br-oh, r-n-boc-4-bromophenylalanine, r-3-4-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-4-br-phe-oh, boc-d-4-bromophe, boc-4'-bromo-d-phe, boc-p-bromo-d-phe-oh, n-boc-4-bromo-d-phenylalanine
PubChem CID 7020327
IUPAC Name (2R)-3-(4-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)Br)C(=O)O

Specifications

Melting Point 119°C
Color White
Quantity 1 g
Formula Weight 344.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-4-bromo-D-phenylalanine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.