Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-D-glutamic Acid 97.0+%, TCI America™
SDP

Catalog No. B298725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B298725G 25 g
B29875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B298725G Supplier TCI America Supplier No. B298725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34404-28-9
Molecular Formula C10H17NO6
Molecular Weight (g/mol) 247.247
MDL Number MFCD00190790
InChI Key AQTUACKQXJNHFQ-ZCFIWIBFSA-N
Synonym boc-d-glu-oh, boc-d-glutamic acid, n-tert-butoxycarbonyl-d-glutamic acid, r-2-tert-butoxycarbonyl amino pentanedioic acid, d-glutamic acid, n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino pentanedioic acid, pubchem20973, n-boc-d-glutamic acid, ksc493m2t, 2r-2-2-methylpropan-2-yl oxycarbonylamino pentanedioic acid
PubChem CID 7019046
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioic acid
SMILES CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)O

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 25 g
Formula Weight 247.25
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-glutamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.