Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)ethanolamine, 98%

Catalog No. p-78426
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass Bottle

CAS: 26690-80-2 | C7H15NO3 | 161.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26690-80-2
Molecular Formula C7H15NO3
Molecular Weight (g/mol) 161.2
MDL Number MFCD00056657
InChI Key GPTXCAZYUMDUMN-UHFFFAOYSA-N
Synonym n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester
PubChem CID 2733206
IUPAC Name tert-butyl N-(2-hydroxyethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4485 to 1.4505
Linear Formula HOCH2CH2NHCO2C(CH3)3
Density 1.0420g/mL
Quantity 5 mL
Flash Point >110°C
Formula Weight 161.2
Percent Purity 98%
Specific Gravity 1.042
Physical Form Viscous Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)ethanolamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.