Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-L-phenylalanine Methyl Ester 98.0+%, TCI America™
SDP

Catalog No. B425925G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 51987-73-6
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.34
MDL Number MFCD00076977
InChI Key SDSWSVBXRBXPRL-UHFFFAOYNA-N
Synonym boc-phe-ome, boc-l-phenylalanine methyl ester, l-boc-phenyl-alanine methyl ester, n-boc-l-phenylalanine methyl ester, n-tert-butoxycarbonyl-l-phenylalanine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-phenylpropanoate, s-2-tert-butoxycarbonylamino-3-phenyl-propionic acid methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-phenylpropanoate, t-boc-l-phe-ome, pubchem13507
PubChem CID 9882138
IUPAC Name methyl 2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate
SMILES COC(=O)C(CC1=CC=CC=C1)NC(=O)OC(C)(C)C

Specifications

Melting Point 39°C
Color White
Quantity 25 g
Formula Weight 279.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-phenylalanine Methyl Ester
EINECSNumber (3)-4367
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.