Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-tert-Butoxycarbonyl-L-prolinol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
69610-40-8
C10H19NO3
201.26
MFCD00066232
BFFLLBPMZCIGRM-QMMMGPOBSA-N
n-boc-l-prolinol,boc-l-prolinol,s-1-boc-2-pyrrolidinemethanol,s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate,tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate,1-boc-l-prolinol,n-tert-butoxycarbonyl-l-prolinol,boc-prolinol,s---1-boc-2-pyrrolidinemethanol,boc-pro-ol
643448
tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC1CO
69610-40-8
C10H19NO3
201.26
MFCD00066232
BFFLLBPMZCIGRM-QMMMGPOBSA-N
n-boc-l-prolinol,boc-l-prolinol,s-1-boc-2-pyrrolidinemethanol,s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate,tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate,1-boc-l-prolinol,n-tert-butoxycarbonyl-l-prolinol,boc-prolinol,s---1-boc-2-pyrrolidinemethanol,boc-pro-ol
643448
tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC1CO
Melting Point 60°C to 64°C
Specific Rotation -52
Color White
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: insoluble in water
Formula Weight 201.26
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material N-tert-Butoxycarbonyl-L-prolinol
Show More Show Less