Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nickel(II) sulfate hexahydrate, 98+%, ACS reagent
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 53.0°C |
| Color | Blue-Green |
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Physical Form | Crystals |
| Quantity | 1 kg |
| Assay Percent Range | 98.0 to 102% (Complexometry) |
| Linear Formula | NiSO4·6H2O |
| Merck Index | 15,6602 |
| Solubility Information | Solubility in water: 625g/L (20°C). Other solubilities: sparingly soluble in alcohol |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Very toxic to aquatic life with long lasting effects. Harmful if inhaled. Suspected of causing genetic defects. Causes damage to organs through prolonged or repeated exposure. Causes skin irritation. M |
| Precautionary Statement | GHS P Statement Do not breathe dust/fume/gas/mist/vapors/spray. IF exposed or concerned: Get medical advice/attention. Wear protective gloves/protective clothing/eye protection/face protection. Avoid release to the environment. |
| RTECSNumber | QR9600000 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction