Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-tert-Butylglycine hydrochloride, 97%

Catalog No. AAH6427006
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6427006 5 g
AAH6427003 1 g
AAH6427014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6427006 Supplier Thermo Scientific Chemicals Supplier No. H6427006
Add to Cart

CAS: 6939-23-7 | C6H14ClNO2 | 167.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6939-23-7
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.63
MDL Number MFCD03788441
InChI Key QORBVLNEGUIPND-UHFFFAOYSA-N
Synonym 2-tert-butylamino acetic acid hydrochloride, n-tert-butylglycine hydrochloride, n-t-butylglycine hydrochloride, n-t-butylglycine hcl, unii-b8y856vh2e, t-butylglycine hydrochloride, glycine, n-1,1-dimethylethyl-, hydrochloride 1:1, glycine, n-1,1-dimethylethyl-, hydrochloride, t-butylaminoacetic acid hydrochloride, glycine, n-tert-butyl-, hydrochloride
PubChem CID 17748195
IUPAC Name hydrogen 2-(tert-butylamino)acetic acid chloride
SMILES [H+].[Cl-].CC(C)(C)NCC(O)=O

Specifications

Melting Point 223°C to 224°C
Color White
Linear Formula (CH3)3CNHCH2COOH·HCl
Beilstein 6296616
Quantity 5 g
Formula Weight 167.63
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-tert-Butylglycine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.