Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-N-Tosyl-L-glutamic acid, 98%, Thermo Scientific™

Catalog No. AC295151000
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4816-80-2
Molecular Formula C12H15NO6S
Molecular Weight (g/mol) 301.31
MDL Number MFCD00020545
InChI Key KKOZKXBAPIYWAT-UHFFFAOYNA-N
Synonym s-2-4-methylphenylsulfonamido pentanedioic acid, tosyl-l-glutamic acid, n-tosyl-l-glutamic acid, n-p-tolylsulphonyl-l-glutamic acid, +-n-tosyl-l-glutamic acid, 2s-2-4-methylbenzenesulfonamido pentanedioic acid, l-glutamic acid, n-4-methylphenyl sulfonyl, tos-glu-oh, 2-4-methylphenylsulfonamido pentanedioic acid, ksc493e0f
PubChem CID 2063452
IUPAC Name 2-(4-methylbenzenesulfonamido)pentanedioic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(CCC(O)=O)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Beilstein 11, IV, 430
Specific Rotation Condition + 13.00 (20.00°C c=5,acetone)
Specific Rotation +12° to +14° (20°C, 589nm) (c=5, acetone)
Formula Weight 301.32
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material (+)-N-Tosyl-L-glutamic acid, 98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.