Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Trimethylsilyl)dimethylamine, 95%

Catalog No. p-7023215
Click to view available options
Quantity:
10 g
50 g

CAS: 2083-91-2 | C5H15NSi | 117.27 g/mol

N,N-Dimethyltrimethylsilylamine was used in the synthesis of phosphoramidite. It was also employed as a reagent in the preparation of iminium salts and amides as well as for the silylation of polymers. A combination of N-(trimethylsilyl)dimethylamine and MeI was also effective to give p-methylbenzoic acid with a 85% yield based on 90% conversion from the corresponding Me ester. DETA has been treated with N-(trimethylsilyl)-dimethylamine to yield mono-, di- and tri- silylated derivatives

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyltrimethylsilylamine was used in the synthesis of phosphoramidite. It was also employed as a reagent in the preparation of iminium salts and amides as well as for the silylation of polymers. A combination of N-(trimethylsilyl)dimethylamine and MeI was also effective to give p-methylbenzoic acid with a 85% yield based on 90% conversion from the corresponding Me ester. DETA has been treated with N-(trimethylsilyl)-dimethylamine to yield mono-, di- and tri- silylated derivatives

Solubility
Hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2083-91-2
Molecular Formula C5H15NSi
Molecular Weight (g/mol) 117.27
MDL Number MFCD00008297
InChI Key KAHVZNKZQFSBFW-UHFFFAOYSA-N
Synonym n,n-dimethyltrimethylsilylamine, n,n-dimethylaminotrimethylsilane, silanamine, pentamethyl, dimethylamino trimethylsilane, pentamethylsilylamine, n-trimethylsilyl dimethylamine, tmsdma, dimethylaminotrimethylsilane, silanamine, n,n,1,1,1-pentamethyl, n,n-dimethyltrimethylsilamine
PubChem CID 74965
IUPAC Name N-methyl-N-trimethylsilylmethanamine
SMILES CN(C)[Si](C)(C)C

Specifications

Boiling Point 86°C to 87°C
Quantity 10 g
Linear Formula (CH3)3SiN(CH3)2
UN Number UN2733
Beilstein 1731861
Flash Point −19°C (−3°F)
Solubility Information Hydrolyzes in water.
Formula Weight 117.27
Percent Purity 95%
Odor Amine-like
Refractive Index 1.397
Sensitivity Moisture sensitive
Density 0.732
Chemical Name or Material N-(Trimethylsilyl)dimethylamine
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 218-222-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.