Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic Acid 98.0+%, TCI America™
SDP

Catalog No. T0683250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0683250G 250 g
T068325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0683250G Supplier TCI America Supplier No. T0683250G
Add to Cart
Add to Cart

[Good's buffer component for biological research]

Chemical Identifiers

CAS 7365-44-8
Molecular Formula C6H15NO6S
Molecular Weight (g/mol) 229.247
MDL Number MFCD00007532
InChI Key JOCBASBOOFNAJA-UHFFFAOYSA-N
Synonym tes, n-tris hydroxymethyl methyl-2-aminoethanesulfonic acid, tes buffer, tes free acid, ethanesulfonic acid, 2-2-hydroxy-1,1-bis hydroxymethyl ethyl amino, 2-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino ethanesulfonic acid, 2-2-hydroxy-1,1-dihydroxymethyl-ethylamino-ethanesulfonic acid, tris hydroxymethyl methyl-2-aminomethane sulfonic acid, 2-tris hydroxymethyl methylamino ethane-1-sulphonic acid, n-tri hydroxymethyl methyltaurine
PubChem CID 81831
ChEBI CHEBI:44356
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid
SMILES C(CS(=O)(=O)O)NC(CO)(CO)CO

Specifications

Melting Point 216°C
Color White
Quantity 250 g
Formula Weight 229.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Tris(hydroxymethyl)methyl-2-aminoethanesulfonic Acid [Good′s buffer component for biological
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.