Learn More
N-[Tris(hydroxymethyl)methyl]acrylamide, 99%
Description
N-[Tris(hydroxymethyl)methyl] acrylamide is used to prepare N-(5-hydroxymethyl-2-phenyl-[1,3]dioxan-5-yl)-acrylamide by reaction with benzaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-[Tris(hydroxymethyl)methyl] acrylamide is used to prepare N-(5-hydroxymethyl-2-phenyl-[1,3]dioxan-5-yl)-acrylamide by reaction with benzaldehyde.
Solubility
Soluble in water.
Notes
Hygroscopic. Light sensitive. Incompatible with strong oxdizing agents.

Specifications
Specifications
Melting Point | 134°C to 138°C |
Linear Formula | CH2=CHCONHC(CH2OH)3 |
Quantity | 25 g |
Sensitivity | Hygroscopic; Light sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 175.18 |
Percent Purity | 99% |
Chemical Name or Material | N-[Tris(hydroxymethyl)methyl]acrylamide |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 237-645-4 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.