Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Valerylglycine Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. p-7214093
Click to view available options
Quantity:
10 mg
50 mg
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 24003-66-5
Molecular Formula C7H13NO3
Linear Formula C7H13NO3
Quantity 250 mg
MDL Number MFCD01910091
UN Number NONH for all modes of transport
Synonym 2-(Pentanoylamino)acetic acid; N-(1-Oxopentyl)glycine; N-Pentanoylglycine; Valeryl glycine
Formula Weight 159.18
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.