Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Vinylcarbazole, 97%

Catalog No. p-3593481
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 1484-13-5 | C14H11N | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1484-13-5
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00004966,MFCD00134336
InChI Key KKFHAJHLJHVUDM-UHFFFAOYSA-N
Synonym 9-vinylcarbazole, n-vinylcarbazole, 9-vinyl-9h-carbazole, 9h-carbazole, 9-ethenyl, n-vinyl carbazole, vinylcarbazole, 1-vinylcarbazole, poly 9-vinylcarbazole, carbazole, 9-vinyl, poly-n-vinylcarbazole
PubChem CID 15143
IUPAC Name 9-ethenylcarbazole
SMILES C=CN1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 63°C to 65°C
Color White to Yellow
Boiling Point 154°C (3.0 mmHg)
Flash Point 182°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 193.24
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material N-Vinylcarbazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause an allergic skin reaction.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-055-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.