Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-WASP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323279
Description
N-WASP Polyclonal antibody specifically detects N-WASP in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
N-WASP | |
Polyclonal | |
Western Blot 1:1000 | |
N-WASPneural WiskNWASP, WiskDKFZp779G0847, WiskMGC48327 | |
N-WASP synthetic peptide (coupled to KLH) corresponding to amino acid residues in the N-terminal region of human N-WASP. This N-WASP peptide sequence is 100% homologous to rat and mouse N-WASP, and has low homology to the corresponding region in the human WASP. [UniProt# O00401] | |
0.1 mL | |
Phospho Specific | |
8976 | |
Store at -20°C. | |
IgG |
Western Blot | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction