Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ N-WASP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5105714
Description
Antibody detects endogenous levels of total N-WASP.
The Wiskott-Aldrich syndrome (WAS) family of proteins share similar domain structure, and are involved in transduction of signals from receptors on the cell surface to the actin cytoskeleton. The presence of a number of different motifs suggests that they are regulated by a number of different stimuli, and interact with multiple proteins. Recent studies have demonstrated that these proteins, directly or indirectly, associate with the small GTPase, Cdc42, known to regulate formation of actin filaments, and the cytoskeletal organizing complex, Arp2/3. The WASL gene product is a homolog of WAS protein, however, unlike the latter, it is ubiquitously expressed and shows highest expression in neural tissues. It has been shown to bind Cdc42 directly, and induce formation of long actin microspikes.
Specifications
| N-WASP | |
| Polyclonal | |
| Unconjugated | |
| WASL | |
| 2900021I12Rik; 3110031I02Rik; DKFZp779G0847; MGC48327; n wasp; neural Wiskott-Aldrich syndrome protein; NWASP; N-WASP; TRS4; WASL; WASP like actin nucleation promoting factor; WASPB; Wiskott-Aldrich syndrome like; Wiskott-Aldrich syndrome-like; Wiskott-Aldrich syndrome-like (human) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 73178, 8976 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00401, Q91YD9 | |
| WASL | |
| A synthesized peptide derived from human WASL(Accession O00401), corresponding to amino acid residues E231-P281. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction