Learn More
Invitrogen™ N-WASP Recombinant Rabbit Monoclonal Antibody (K01_1W05)

Rabbit Recombinant Monoclonal Antibody
$312.00 - $544.00
Specifications
| Antigen | N-WASP |
|---|---|
| Clone | K01_1W05 |
| Concentration | 1 mg/mL |
| Content And Storage | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Description
N-WASP Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot
The Wiskott-Aldrich syndrome (WAS) family of proteins share similar domain structure, and are involved in transduction of signals from receptors on the cell surface to the actin cytoskeleton. The presence of a number of different motifs suggests that they are regulated by a number of different stimuli, and interact with multiple proteins. Recent studies have demonstrated that these proteins, directly or indirectly, associate with the small GTPase, Cdc42, known to regulate formation of actin filaments, and the cytoskeletal organizing complex, Arp2/3. The WASL gene product is a homolog of WAS protein, however, unlike the latter, it is ubiquitously expressed and shows highest expression in neural tissues. It has been shown to bind Cdc42 directly, and induce formation of long actin microspikes.Specifications
| N-WASP | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4 | |
| O00401, Q91YD9 | |
| 73178, 8976 | |
| A synthetic peptide of human N-WASP. | |
| Antibody |
| K01_1W05 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| WASL | |
| 2900021I12Rik; 3110031I02Rik; DKFZp779G0847; MGC48327; n wasp; neural Wiskott-Aldrich syndrome protein; NWASP; N-WASP; TRS4; WASL; WASP like actin nucleation promoting factor; WASPB; Wiskott-Aldrich syndrome like; Wiskott-Aldrich syndrome-like; Wiskott-Aldrich syndrome-like (human) | |
| WASL | |
| Primary | |
| Protein A/G |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.