Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Noroxycodone-D3 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco N0321ML
Description
This stable-labeled certified reference solution is suitable for quantitation of noroxycodone levels in urine, serum, or plasma by LC/MS or GC/MS for applications such as urine drug testing, clinical toxicology, forensic analysis, pain prescription monitoring, or isotope dilution methods. Noroxycodone is a major urinary metabolite of oxycodone, an opiate analgesic drug used for the management of moderate to severe pain, both acute and chronic.
Specifications
| 67-56-1 | |
| MFCD08272692 | |
| 100 μg/mL in methanol (as free base) | |
| 340.82 | |
| 1 mL |
| C17H16D3NO4·HCl | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| Noroxycodone-D3 hydrochloride solution | |
| Ampule | |
| 9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Recommended Storage : -20°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?Share a Content Correction