Learn More
CERILLIANT™ Naloxone-3ß-D-glucuronide Solution, 1.0 mg/mL in Methanol:Water (9:1), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco N0991ML
Description
Naloxone-3β-D-glucuronide is a major urinary metabolite of naloxone, an opiate drug used to counter the effects of opioid overdose. This certified reference solution is suitable as starting material for preparation of calibrators and controls in naloxone testing by LC/MS or GC/MS for forensic analysis, urine drug testing, or clinical toxicology applications.
Specifications
| 67-56-1 | |
| UN 1230 - class 3 - PG 2 - Methanol, solution | |
| 503.50 | |
| 1 mL |
| C25H29NO10 | |
| 1.0 mg/mL in methanol:water (9:1) | |
| Ampule |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Recommended Storage : -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
CERILLIANT is a registered trademark of Cerilliant Corporation