Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N1,N1-Dimethyl-1-phenyl-1,2-ethanediamine, 98%

Catalog No. p-7033223
Click to view available options
Quantity:
250 mg
1 g

CAS: 6342-21-8 | C10H16N2 | 164.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6342-21-8
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.252
MDL Number MFCD01631934
InChI Key NFSAPTWLWWYADB-UHFFFAOYSA-N
Synonym 2-amino-1-phenylethyl dimethylamine, n1,n1-dimethyl-1-phenyl-1,2-ethanediamine, n1,n1-dimethyl-1-phenylethane-1,2-diamine, n*1*,n*1*-dimethyl-1-phenyl-ethane-1,2-diamine, n∼1∼,n∼1∼-dimethyl-1-phenylethane-1,2-diamine, 1,2-ethanediamine, n1,n1-dimethyl-1-phenyl, acmc-20apq4, daa-0-0, n,n-dimethyl-1-phenyl-ethane-1,2-diamine
PubChem CID 241652
IUPAC Name N,N-dimethyl-1-phenylethane-1,2-diamine
SMILES CN(C)C(CN)C1=CC=CC=C1

Specifications

Quantity 250 mg
UN Number UN2735
Formula Weight 164.25
Percent Purity 98%
Chemical Name or Material N1,N1-Dimethyl-1-phenyl-1,2-ethanediamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.