Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N2-Isobutyrylguanosine Monohydrate, TCI America™
SDP

Catalog No. I0699100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0699100MG 100 mg
I06991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0699100MG Supplier TCI America Supplier No. I0699100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64350-24-9
Molecular Formula C14H21N5O7
Molecular Weight (g/mol) 371.35
MDL Number MFCD00274102,MFCD00274102
InChI Key JXIMAIWXKDBJKP-UHFFFAOYNA-N
PubChem CID 56924510
IUPAC Name N-{9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6-oxo-6,9-dihydro-3H-purin-2-yl}-2-methylpropanamide hydrate
SMILES O.CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C1OC(CO)C(O)C1O

Specifications

Melting Point 148°C
Color White
Quantity 100 mg
Formula Weight 353.34
Physical Form Crystalline Powder
Chemical Name or Material N2-Isobutyrylguanosine Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.