Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N2-Methyl-L-arginine, Tocris Bioscience™

Catalog No. p-4817498
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Lysosomal system c L-arginine uptake inhibitor

Selective L-arginine uptake inhibitor (Ki = 500 μM). Inhibits uptake of arginine by the lysosomal system c in human fibroblast in vitro.

Chemical Identifiers

CAS 2480-28-6
Molecular Formula C7H16N4O2
Molecular Weight (g/mol) 188.231
InChI Key NTWVQPHTOUKMDI-YFKPBYRVSA-N
Synonym n-methyl-l-arginine, n2-methyl-l-arginine, arginine, n2-methyl-, l, unii-88rbj4ze9r, 88rbj4ze9r, l-monomethyl-l-arginine, s-5-guanidino-2-methylamino pentanoic acid, n 2-methyl-l-arginine, n∼5∼-diaminomethylene-n∼2∼-methylornithine, n-mearg
PubChem CID 73046
IUPAC Name (2S)-5-(diaminomethylideneamino)-2-(methylamino)pentanoic acid
SMILES CNC(CCCN=C(N)N)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 188.23
Percent Purity >98%
Chemical Name or Material N2-Methyl-L-arginine
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.