Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N2-(tert-Butoxycarbonyl)-4-fluoro-1,2-phenylenediamine 98.0+%, TCI America™
SDP

Catalog No. B42371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42371G 1 g
B4237200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42371G Supplier TCI America Supplier No. B42371G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 362670-07-3
Molecular Formula C11H15FN2O2
Molecular Weight (g/mol) 226.25
MDL Number MFCD07787180
InChI Key FELBQUPQCJZQQX-UHFFFAOYSA-N
Synonym N2-Boc-4-fluoro-1,2-phenylenediamine, tert-Butyl (2-Amino-5-fluorophenyl)carbamate, (2-Amino-5-fluorophenyl)carbamic Acid tert-Butyl Ester
PubChem CID 53420357
IUPAC Name tert-butyl N-(2-amino-5-fluorophenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=C(N)C=CC(F)=C1

Specifications

Melting Point 101°C
Color Gray-Red
Quantity 1 g
Formula Weight 226.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N2-(tert-Butoxycarbonyl)-4-fluoro-1,2-phenylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.