Learn More
Invitrogen™ N2DL4 Monoclonal Antibody (709116)

Description
In Western blots, approximately 20% cross-reactivity with recombinant human (rh) ULBP-2 is observed and no cross-reactivity with rhULBP-1 or rhULBP-3 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | N2DL4 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 709116 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | RAET1E |
| Gene Accession No. | Q8TD07 |
| Gene Alias | bA350J20.7; LETAL; Lymphocyte effector toxicity activation ligand; MGC125309; N2DL4; N2DL-4; NKG2D ligand 4; NKG2DL4; Rae-1 epsilon; RAE1 like transcript 4; RAE1epsilon; RAE-1-epsilon; RAE-1-like transcript 4; RAET1E; RAET1E2; Retinoic acid early transcript 1E; retinoic acid early-inducible protein 1-epsilon; RL-4; UL16 binding protein 4; UL16-binding protein 4; ULBP 4; ULBP4; ULBP4MGC125308; UNQ1867/PRO4303 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.