Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ N4BP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117135
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117135 Supplier Invitrogen™ Supplier No. PA5117135
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Jurkat, 293T, Mouse brain, Mouse pancreas Immunogen sequence: TQGEESLFET DDDVITVGDV RPPEYTQSAA TGPSSARDEV VVQEDSRNKA RTPVSELTKH MDTVFSSSPD VLFVPVNGLS PDEDALSKDR VCHKRRSSDT EERHTKKQFS LENVPEGELL PDGKGSAGNE VIDLSDPASN STNLSPDGKD TTEEMEYNIL VNFFKTMGYS QEIVEKVIRE YGPSTEPLLL LEEIEKENKR LQEDRDFPPC TVYPDASQSR NAGVGSTTNE LTADSTPKKA QSHTEQSMVE RFSQLPFKDS KHCTSNCKVN SFRTVPVGQK QEIWGSKQNS SCTVDLETDG HSASAASASP KDISFVSRGA SGHQQRNPAF PENGFQQQTE PLLPNNTKPA CEKRSGSCSS PQPKPNYPPL SPPLPLPQLL PSVTEARLGG SSDHIDSSVT G.

Nedd4-binding partner-1 (N4BP1) is a developmentally expressed protein interactor and monoubiquitylation substrate of Nedd4, a HECT domain-containing ubiquitin ligase that mediates ubiquitylation and proteasome degradation of target proteins and may have a fundamental role to play in embryonic processes. N4BP1 can undergo Nedd4-mediated polyubiquitylation and proteosomal degradation. N4BP1 also functions as a negative regulator of AIF4 (also known as ITCH or Atrophin-1), an E3-ubiquitin ligase, by interacting with the second WW domain of AIF4 and impairing ubiquitination of substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen N4BP1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene N4BP1
Gene Accession No. O75113, Q6A037
Gene Alias AI481586; C81621; Kiaa0615; N4BP1; Nedd4 binding protein 1; NEDD4-binding protein 1; RGD1305179
Gene Symbols N4BP1
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80750, 9683
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.