Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N4BP2 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298059AF647
Description
N4BP2 Polyclonal specifically detects N4BP2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
N4BP2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
B3BPKIAA1413, BCL-3-binding protein, EC 3.-, FLJ10680, NEDD4 binding protein 2, NEDD4-binding protein 2 | |
Produced in rabbits immunized with E. coli-derived Human N4BP2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
55728 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction