Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ N4BP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565335
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565335 Supplier Invitrogen™ Supplier No. PA565335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSSDSLAQREH RSRMPKTGLS EPNLEIGTND KMNEISLSTA HEACWGTSSQ KLKTLGSSNL GSSEMLLSEM TCESQTCLSK KSHGQHT Highest antigen sequence identity to the following orthologs - mouse 60%, rat 69%.

Has 5'-polynucleotide kinase and nicking endonuclease activity. May play a role in DNA repair or recombination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen N4BP2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene N4BP2
Gene Accession No. Q86UW6
Gene Alias B3BP; BCL-3-binding protein; KIAA1413; N4BP2; NEDD4 binding protein 2; NEDD4-binding protein 2
Gene Symbols N4BP2
Host Species Rabbit
Immunogen Recombinant Human N4BP2. Recombinant protein control fragment (Product #RP-106030).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55728
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.