Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ N4BP2L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110009
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110009 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110009 Supplier Invitrogen™ Supplier No. PA5110009
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FRKHLYLLRG LPGSGKTTLA RQLQHDFPRA LIFSTDDFFF REDGAYEFNP DFLEEAHEWN QKRARKAMRN GISPIIIDNT N.

N4BP2L1 (NEDD4 Binding Protein 2 Like 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includekinase activity. An important paralog of this gene isN4BP2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen N4BP2L1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene N4BP2L1
Gene Accession No. Q3V2Q8
Gene Alias 2410024N18Rik; AI428195; B230342M21Rik; CG018; CG081; hypothetical gene CG018; N4BP2L1; NEDD4 binding protein 2 like 1; NEDD4 binding protein 2-like 1; NEDD4-binding protein 2-like 1
Gene Symbols N4BP2L1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 40-120 of human N4BP2L1 (NP_001073159).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100637
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.