Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ N4BP2L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558616
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558616 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558616 Supplier Invitrogen™ Supplier No. PA558616
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KIHRMKERYE HDVTFHSVLH AEKPSRMNRN QDRNNALPSN NARYWNSYTE FPNRRAHGGF TNESSYHRRG Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 69%.

N4BP2L1 (NEDD4 Binding Protein 2 Like 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includekinase activity. An important paralog of this gene isN4BP2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen N4BP2L1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene N4BP2L1
Gene Accession No. Q5TBK1
Gene Alias 2410024N18Rik; AI428195; B230342M21Rik; CG018; CG081; hypothetical gene CG018; N4BP2L1; NEDD4 binding protein 2 like 1; NEDD4 binding protein 2-like 1; NEDD4-binding protein 2-like 1
Gene Symbols N4BP2L1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human N4BP2L1. Recombinant protein control fragment (Product #RP-97968).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90634
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.